Assay Details
Target Gene Details
Entrez Gene ID: | 85445 |
Gene Name: | contactin associated protein like 4 |
Gene Aliases: |
CASPR4 |
Location: |
Chr.16:76277217-76563610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTNAP4 | NM_001322178.1 | NP_001309107.1 | ||
| NM_001322179.1 | NP_001309108.1 | |||
| NM_001322180.1 | NP_001309109.1 | |||
| NM_001322181.1 | NP_001309110.1 | |||
| NM_001322187.1 | NP_001309116.1 | |||
| NM_001322188.1 | NP_001309117.1 | |||
| NM_001322189.1 | NP_001309118.1 | |||
| NM_001322190.1 | NP_001309119.1 | |||
| NM_001322191.1 | NP_001309120.1 | |||
| NM_033401.4 | NP_207837.2 | |||
| NM_138994.4 | NP_620481.2 | |||
| NR_136210.1 | ||||
| NR_136213.1 | ||||
| NR_136216.1 | ||||
| XM_011523403.2 | XP_011521705.1 | |||
| XM_017023805.1 | XP_016879294.1 | |||
| AB051550.2 | ||||
| AB100092.1 | BAC55270.1 | |||
| AB100094.1 | BAC55272.1 | |||
| AF463518.1 | 1 | 157 | AAL68839.1 | |
| AK057327.1 | BAB71429.1 | |||
| DB050637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065644 | Chr.16:76132681 - 76441737 on Build GRCh38 | Gain |
|
| esv3892895 | Chr.16:76153094 - 76478682 on Build GRCh38 | Gain |
|
| nsv1064607 | Chr.16:75753231 - 76415204 on Build GRCh38 | Gain |
|
| nsv1059162 | Chr.16:75895450 - 76696919 on Build GRCh38 | Gain |
|
| dgv5192n54 | Chr.16:75901127 - 76701352 on Build GRCh38 | Gain |
|
| esv2763142 | Chr.16:76237762 - 76427944 on Build GRCh38 | Loss |
|
| nsv833286 | Chr.16:76209832 - 76410386 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map