Assay Details
Target Gene Details
Entrez Gene ID: | 221477 |
Gene Name: | chromosome 6 open reading frame 89 |
Gene Aliases: |
BRAP |
Location: |
Chr.6:36834886-36928964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C6orf89 | XM_017010434.1 | XP_016865923.1 |
Target Gene Details
Entrez Gene ID: | 51645 |
Gene Name: | peptidylprolyl isomerase like 1 |
Gene Aliases: |
CGI-124, CYPL1, PPIase, hCyPX |
Location: |
Chr.6:36854829-36875024 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL1 | NM_016059.4 | NP_057143.1 | ||
| AF151882.1 | AAD34119.1 | |||
| AK026636.1 | ||||
| AY359032.1 | AAQ89391.1 | |||
| BC003048.1 | AAH03048.1 | |||
| BU195579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map