Assay Details
Target Gene Details
Entrez Gene ID: | 130752 |
Gene Name: | malate dehydrogenase 1B |
Gene Aliases: |
RP11-95H11 |
Location: |
Chr.2:206737763-206765549 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDH1B | NM_001039845.2 | 12 | 2472 | NP_001034934.1 |
| NM_001282940.1 | 12 | 2469 | NP_001269869.1 | |
| NR_104261.1 | 10 | 2111 | ||
| XM_005246315.4 | XP_005246372.1 | |||
| XM_011510628.2 | XP_011508930.1 | |||
| XM_017003373.1 | XP_016858862.1 | |||
| XM_017003374.1 | XP_016858863.1 | |||
| AI076012.1 | 1 | 93 | ||
| AK302250.1 | ||||
| BC033509.1 | AAH33509.1 | |||
| BF979825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011347 | Chr.2:206384007 - 207085773 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map