Assay Details
Target Gene Details
Entrez Gene ID: | 91452 |
Gene Name: | acyl-CoA binding domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.10:27182624-27248674 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACBD5 | XM_006717528.3 | XP_006717591.3 | ||
| XM_017016884.1 | XP_016872373.1 | |||
| XM_017016885.1 | XP_016872374.1 | |||
| XM_017016886.1 | XP_016872375.1 | |||
| XM_017016890.1 | XP_016872379.1 | |||
| XM_017016893.1 | XP_016872382.1 | |||
| XM_017016894.1 | XP_016872383.1 |
Target Gene Details
Entrez Gene ID: | 387646 |
Gene Name: | leucine rich repeat containing 37 member A6, pseudogene |
Gene Aliases: |
LRRC37E |
Location: |
Chr.10:27245853-27252306 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC37A6P | NR_003525.2 | 1 | 3610 | |
| AK126872.1 | 1 | 1866 | BAC86731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622711 | Chr.10:27242765 - 27300671 on Build GRCh38 | Gain |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
|
| nsv8614 | Chr.10:27215558 - 27539477 on Build GRCh38 | Loss |
|
| nsv947786 | Chr.10:27246212 - 27252983 on Build GRCh38 | Duplication |
|
| esv2422253 | Chr.10:27142437 - 27286441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map