Assay Details
Target Gene Details
Entrez Gene ID: | 54093 |
Gene Name: | SET domain containing 4 |
Gene Aliases: |
C21orf18, C21orf27 |
Location: |
Chr.21:36034541-36060818 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD4 | NM_001007259.2 | 7 | 992 | NP_001007260.1 |
| NM_001007261.2 | 8 | 1086 | NP_001007262.1 | |
| NM_001286752.1 | 8 | 1475 | NP_001273681.1 | |
| NM_017438.4 | 7 | 1332 | NP_059134.1 | |
| NR_040087.2 | 6 | 1243 | ||
| XM_011529636.1 | 7 | 1381 | XP_011527938.1 | |
| XM_011529637.1 | 7 | 1446 | XP_011527939.1 | |
| XM_011529638.2 | 7 | 1354 | XP_011527940.1 | |
| XM_011529639.1 | 7 | 1132 | XP_011527941.1 | |
| XM_011529640.2 | 7 | 1397 | XP_011527942.1 | |
| XM_011529642.1 | 8 | 1475 | XP_011527944.1 | |
| XM_011529643.1 | 8 | 1426 | XP_011527945.1 | |
| XM_011529644.1 | 8 | 987 | XP_011527946.1 | |
| XM_017028403.1 | 7 | 923 | XP_016883892.1 | |
| XM_017028404.1 | 8 | 1031 | XP_016883893.1 | |
| XM_017028405.1 | 7 | 1332 | XP_016883894.1 | |
| AB004848.1 | 8 | 1039 | ||
| AB004849.1 | 2 | 542 | ||
| AB004852.1 | 6 | 863 | ||
| AB004853.1 | 8 | 1035 | ||
| AF391112.1 | 6 | 810 | AAL34503.1 | |
| AK001660.1 | 7 | 899 | BAA91819.1 | |
| AK095215.1 | 6 | 2199 | ||
| AK300009.1 | 6 | 943 | ||
| BC002898.2 | 7 | 931 | AAH02898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470899 | Chr.21:36001424 - 36096978 on Build GRCh38 | Loss |
|
| nsv459247 | Chr.21:36003345 - 36087866 on Build GRCh38 | Loss |
|
| nsv964547 | Chr.21:36003423 - 36090261 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map