Assay Details
Target Gene Details
Entrez Gene ID: | 55125 |
Gene Name: | centrosomal protein 192 |
Gene Aliases: |
PPP1R62 |
Location: |
Chr.18:12991307-13125053 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP192 | NM_032142.3 | 8 | 934 | NP_115518.3 |
| XM_005258107.3 | 8 | 989 | XP_005258164.1 | |
| XM_005258109.3 | 8 | 989 | XP_005258166.1 | |
| XM_006722326.3 | 8 | 989 | XP_006722389.1 | |
| XM_006722327.3 | 8 | 989 | XP_006722390.1 | |
| XM_006722330.3 | 8 | 989 | XP_006722393.1 | |
| XM_011525673.2 | 8 | 989 | XP_011523975.1 | |
| XM_011525675.2 | 5 | 538 | XP_011523977.1 | |
| XM_017025803.1 | 7 | 824 | XP_016881292.1 | |
| XM_017025804.1 | 8 | 989 | XP_016881293.1 | |
| XM_017025806.1 | 5 | 564 | XP_016881295.1 | |
| XM_017025807.1 | 5 | 564 | XP_016881296.1 | |
| AK096548.1 | 8 | 934 | ||
| BX648232.1 | 5 | 520 | ||
| KJ567064.1 | 7 | 854 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576524 | Chr.18:12918319 - 13183610 on Build GRCh38 | Gain |
|
| nsv1057083 | Chr.18:12916780 - 13207539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map