Assay Details
Target Gene Details
Entrez Gene ID: | 22948 |
Gene Name: | chaperonin containing TCP1 subunit 5 |
Gene Aliases: |
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon |
Location: |
Chr.5:10249921-10266412 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT5 | NM_001306153.1 | NP_001293082.1 | ||
| NM_001306154.1 | NP_001293083.1 | |||
| NM_001306155.1 | NP_001293084.1 | |||
| NM_001306156.1 | NP_001293085.1 | |||
| NM_012073.4 | NP_036205.1 | |||
| AK289353.1 | ||||
| AK290393.1 | ||||
| AK293341.1 | ||||
| AK293437.1 | ||||
| AK301760.1 | ||||
| AK301985.1 | ||||
| AK302368.1 | ||||
| AK302383.1 | ||||
| AK303086.1 | ||||
| AK303147.1 | ||||
| AK316376.1 | ||||
| BC002971.1 | AAH02971.1 | |||
| BC006543.2 | AAH06543.1 | |||
| BC009454.1 | AAH09454.1 | |||
| BC035499.2 | AAH35499.1 | |||
| D43950.2 | ||||
| FJ224303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597112 | Chr.5:10236924 - 10262046 on Build GRCh38 | Gain |
|
| nsv1029129 | Chr.5:10133592 - 10300387 on Build GRCh38 | Gain |
|
| esv33231 | Chr.5:9985609 - 10272597 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map