Assay Details
Target Gene Details
Entrez Gene ID: | 23479 |
Gene Name: | iron-sulfur cluster assembly enzyme |
Gene Aliases: |
2310020H20Rik, HML, ISU2, NIFU, NIFUN, hnifU |
Location: |
Chr.12:108561463-108569384 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISCU | NM_001301140.1 | 6 | 1835 | NP_001288069.1 |
| NM_001301141.1 | 6 | 1846 | NP_001288070.1 | |
| NM_001320042.1 | 5 | 2922 | NP_001306971.1 | |
| NM_014301.4 | 6 | 1845 | NP_055116.1 | |
| NM_213595.3 | 5 | 1749 | NP_998760.1 | |
| NR_135127.1 | 7 | 1945 | ||
| XM_011538100.2 | 6 | 3018 | XP_011536402.1 | |
| XM_011538101.2 | 4 | 1670 | XP_011536403.1 | |
| AK000574.1 | 5 | 592 | ||
| AK022758.1 | 4 | 3346 | ||
| AK057251.1 | 6 | 1846 | ||
| AY009127.1 | 6 | 702 | AAG37427.1 | |
| AY009128.1 | 5 | 606 | AAG37428.1 | |
| BC011906.2 | 5 | 605 | AAH11906.1 | |
| BC061903.1 | 5 | 598 | AAH61903.1 | |
| BG702203.1 | 6 | 686 | ||
| BI597126.1 | ||||
| BM423850.1 | 5 | 595 | ||
| BM921073.1 | 5 | 690 | ||
| BQ002528.1 | 1 | 385 | ||
| BQ020993.1 | 1 | 400 | ||
| BU602914.1 | 5 | 610 | ||
| EU329002.1 | 7 | 802 | ||
| U47101.1 | 4 | 452 | AAC50885.1 |
More Information
Set Membership: |
|

Genomic Map