Assay Details
Target Gene Details
Entrez Gene ID: | 56133 |
Gene Name: | protocadherin beta 2 |
Gene Aliases: |
PCDH-BETA2 |
Location: |
Chr.5:141094607-141097380 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHB2 | NM_018936.3 | 1 | 1153 | NP_061759.1 |
| AB209745.1 | 1 | 929 | BAD92982.1 | |
| AF152495.1 | 1 | 969 | AAD43756.1 | |
| AF217756.1 | 1 | 969 | AAK51623.1 | |
| AK027526.1 | 1 | 1125 | ||
| AK314642.1 | 1 | 1107 | ||
| BC098347.1 | 1 | 1016 | AAH98347.1 | |
| BC098575.1 | 1 | 1117 | AAH98575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map