Assay Details
Target Gene Details
Entrez Gene ID: | 28990 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
HT001 |
Location: |
Chr.3:131013873-131026854 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTE1 | NM_001288950.1 | 3 | 1348 | NP_001275879.1 |
NM_014065.3 | 3 | 1573 | NP_054784.2 | |
NR_110229.1 | 3 | 1573 | ||
XM_017006266.1 | 3 | 1489 | XP_016861755.1 | |
XM_017006267.1 | 3 | 1647 | XP_016861756.1 | |
XM_017006268.1 | 3 | 1568 | XP_016861757.1 | |
XM_017006269.1 | 3 | 1489 | XP_016861758.1 | |
AF113539.1 | 1 | 245 | AAF14876.1 | |
AK027258.1 | 1 | 963 | ||
AK027806.1 | 3 | 1521 | BAB55383.1 | |
AK294156.1 | 3 | 1573 | ||
AK301388.1 | ||||
BC030274.1 | 1 | 193 | ||
BC103709.1 | 1 | 664 | AAI03710.1 | |
BC110616.2 | 3 | 1286 | AAI10617.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
