Assay Details
Target Gene Details
Entrez Gene ID: | 64403 |
Gene Name: | cadherin 24 |
Gene Aliases: |
CDH11L |
Location: |
Chr.14:23047061-23057619 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH24 | NM_022478.3 | 12 | 2070 | NP_071923.2 |
| NM_144985.3 | 11 | 1956 | NP_659422.2 | |
| AK025342.1 | 4 | 374 | BAB15116.1 | |
| AK289576.1 | 11 | 1956 | ||
| AL137477.1 | 3 | 529 | CAB70758.1 | |
| AY260900.1 | 10 | 1789 | AAP20590.1 | |
| AY260901.1 | 11 | 1903 | AAP20591.1 | |
| AY358199.1 | 11 | 1912 | AAQ88566.1 | |
| BC110867.1 | AAI10868.1 | |||
| BX248303.1 | 7 | 980 | CAD62630.1 |
More Information
Set Membership: |
|

Genomic Map