Assay Details
Target Gene Details
Entrez Gene ID: | 7201 |
Gene Name: | thyrotropin releasing hormone receptor |
Gene Aliases: |
TRH-R |
Location: |
Chr.8:109085745-109119785 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRHR | NM_003301.5 | 1 | 63 | NP_003292.1 |
| XM_011517263.2 | 2 | 1201 | XP_011515565.1 | |
| AY493373.1 | AAR84356.1 | |||
| BC105045.1 | 1 | 63 | AAI05046.1 | |
| BC113360.1 | 1 | 63 | ||
| D16845.1 | BAA04120.1 | |||
| X72089.1 | CAA50979.1 | |||
| X75071.1 | CAA52965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1117845 | Chr.8:109058493 - 109089363 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map