Assay Details
Target Gene Details
Entrez Gene ID: | 57379 |
Gene Name: | activation-induced cytidine deaminase |
Gene Aliases: |
AID, ARP2, CDA2, HEL-S-284, HIGM2 |
Location: |
Chr.12:8602166-8612970 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AICDA | NM_020661.2 | 5 | 2624 | NP_065712.1 |
| XM_011520772.2 | 5 | 2713 | XP_011519074.1 | |
| AB040431.1 | 5 | 2621 | BAB12721.1 | |
| EU794587.1 | 5 | 2621 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss |
|
| dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map