Assay Details
Target Gene Details
Entrez Gene ID: | 5286 |
Gene Name: | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha |
Gene Aliases: |
CPK, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2-alpha, PI3K-C2alpha |
Location: |
Chr.11:17086572-17207996 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3C2A | NM_001321378.1 | NP_001308307.1 | ||
| NM_001321380.1 | NP_001308309.1 | |||
| NM_002645.3 | NP_002636.2 | |||
| XM_011520186.2 | XP_011518488.1 | |||
| XM_017017924.1 | XP_016873413.1 | |||
| AK294416.1 | ||||
| AK304262.1 | ||||
| AK309610.1 | ||||
| BC113658.1 | ||||
| BX648778.1 | ||||
| Y13367.1 | CAA73797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| nsv553568 | Chr.11:17006008 - 17299250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map