Assay Details
Target Gene Details
Entrez Gene ID: | 55125 |
Gene Name: | centrosomal protein 192 |
Gene Aliases: |
PPP1R62 |
Location: |
Chr.18:12991307-13125053 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP192 | NM_032142.3 | 11 | 1533 | NP_115518.3 |
| XM_005258107.3 | 11 | 1588 | XP_005258164.1 | |
| XM_005258109.3 | 11 | 1588 | XP_005258166.1 | |
| XM_005258110.1 | 2 | 305 | XP_005258167.1 | |
| XM_006722326.3 | 11 | 1588 | XP_006722389.1 | |
| XM_006722327.3 | 11 | 1588 | XP_006722390.1 | |
| XM_006722330.3 | XP_006722393.1 | |||
| XM_011525673.2 | 11 | 1588 | XP_011523975.1 | |
| XM_011525675.2 | 8 | 1137 | XP_011523977.1 | |
| XM_017025803.1 | 10 | 1423 | XP_016881292.1 | |
| XM_017025804.1 | XP_016881293.1 | |||
| XM_017025805.1 | 2 | 187 | XP_016881294.1 | |
| XM_017025806.1 | 8 | 1163 | XP_016881295.1 | |
| XM_017025807.1 | 8 | 1163 | XP_016881296.1 | |
| AK096548.1 | ||||
| AL833494.2 | 2 | 253 | ||
| BC144480.1 | 2 | 279 | ||
| BC144481.1 | 2 | 279 | ||
| BX648232.1 | 8 | 1119 | ||
| BX649151.1 | 2 | 305 | ||
| KJ567064.1 | 10 | 1453 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576524 | Chr.18:12918319 - 13183610 on Build GRCh38 | Gain |
|
| nsv1057083 | Chr.18:12916780 - 13207539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map