Assay Details
Target Gene Details
Entrez Gene ID: | 100527950 |
Gene Name: | GJA9-MYCBP readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:38862490-38881626 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA9-MYCBP | NR_037633.1 | |||
| NR_037634.1 | ||||
| NR_037635.1 | ||||
| NR_037636.1 | ||||
| NR_037637.1 | ||||
| AK123051.1 | ||||
| FY210901.1 | ||||
| FY211253.1 | ||||
| FY211295.1 |
Target Gene Details
Entrez Gene ID: | 26292 |
Gene Name: | MYC binding protein |
Gene Aliases: |
AMY-1 |
Location: |
Chr.1:38862490-38873378 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCBP | NM_012333.4 | NP_036465.2 | ||
| NR_037632.1 | ||||
| AB007191.2 | BAA22408.1 | |||
| AB451287.1 | ||||
| AB451419.1 | ||||
| AK311886.1 | ||||
| BC008686.1 | AAH08686.1 | |||
| BQ437941.1 | ||||
| BT007109.1 | AAP35773.1 | |||
| BU601195.1 | ||||
| CR456936.1 | CAG33217.1 | |||
| D50692.1 | BAA09338.1 |
More Information
Set Membership: |
|

Genomic Map