Assay Details
Target Gene Details
Entrez Gene ID: | 9841 |
Gene Name: | zinc finger and BTB domain containing 24 |
Gene Aliases: |
BIF1, ICF2, PATZ2, ZNF450 |
Location: |
Chr.6:109462516-109483237 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB24 | NM_014797.2 | 7 | 1648 | NP_055612.2 |
| AB007901.1 | 7 | 1648 | ||
| AK309697.1 | 7 | 1663 | ||
| BC117374.1 | 6 | 1516 | ||
| BC117376.1 | 6 | 1516 |
More Information
Set Membership: |
|

Genomic Map