Assay Details
Target Gene Details
Entrez Gene ID: | 3140 |
Gene Name: | major histocompatibility complex, class I-related |
Gene Aliases: |
HLALS |
Location: |
Chr.1:181033379-181061938 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MR1 | NM_001194999.1 | NP_001181928.1 | ||
| NM_001195000.1 | NP_001181929.1 | |||
| NM_001195035.1 | NP_001181964.1 | |||
| NM_001310213.1 | NP_001297142.1 | |||
| NM_001531.2 | NP_001522.1 | |||
| XM_006711286.3 | XP_006711349.1 | |||
| XM_011509463.2 | XP_011507765.1 | |||
| AF010446.1 | AAD01442.1 | |||
| AF010447.1 | 3 | 627 | AAD01443.1 | |
| AF031469.1 | AAD01933.1 | |||
| AJ249778.1 | CAB77667.1 | |||
| AK222910.1 | BAD96630.1 | |||
| AK290374.1 | ||||
| AK304645.1 | ||||
| BC012485.1 | AAH12485.1 | |||
| BX415555.2 | ||||
| U22963.1 | AAC50174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2719940 | Chr.1:180969855 - 181062846 on Build GRCh38 | Deletion |
|
| nsv946529 | Chr.1:181048790 - 181050587 on Build GRCh38 | Duplication |
|
| esv993520 | Chr.1:181045058 - 181059055 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map