Assay Details
Target Gene Details
Entrez Gene ID: | 128553 |
Gene Name: | teashirt zinc finger homeobox 2 |
Gene Aliases: |
C20orf17, OVC10-2, TSH2, ZABC2, ZNF218 |
Location: |
Chr.20:52972309-53495330 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSHZ2 | NM_173485.5 | NP_775756.3 | ||
| XM_017027640.1 | XP_016883129.1 | |||
| XM_017027641.1 | XP_016883130.1 | |||
| AK091206.1 | BAC03610.1 | |||
| BX640770.1 | CAE45871.1 | |||
| DA647113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459006 | Chr.20:52948734 - 52982503 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map