Assay Details
Target Gene Details
Entrez Gene ID: | 10724 |
Gene Name: | meningioma expressed antigen 5 (hyaluronidase) |
Gene Aliases: |
MEA5, NCOAT, OGA |
Location: |
Chr.10:101784443-101818709 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGEA5 | NM_001142434.1 | NP_001135906.1 | ||
| NM_012215.3 | NP_036347.1 | |||
| XM_017015585.1 | XP_016871074.1 | |||
| XM_017015586.1 | XP_016871075.1 | |||
| AB014579.2 | ||||
| AF036144.2 | AAD05385.2 | |||
| AK000988.1 | 2 | 1395 | ||
| AK002091.1 | ||||
| BC001343.1 | AAH01343.1 | |||
| BC039583.2 | AAH39583.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv171n67 | Chr.10:101769728 - 101817385 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map