Assay Details
Target Gene Details
Entrez Gene ID: | 4361 |
Gene Name: | MRE11 homolog A, double strand break repair nuclease |
Gene Aliases: |
ATLD, HNGS1, MRE11, MRE11B |
Location: |
Chr.11:94415570-94512701 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRE11A | NM_005590.3 | 19 | 3037 | NP_005581.2 |
| NM_005591.3 | 20 | 3014 | NP_005582.1 | |
| XM_005274008.3 | 18 | 2729 | XP_005274065.1 | |
| XM_006718842.3 | 20 | 3118 | XP_006718905.1 | |
| XM_011542837.2 | 20 | 3543 | XP_011541139.1 | |
| XM_017017772.1 | 20 | 3121 | XP_016873261.1 | |
| XM_017017773.1 | 20 | 3011 | XP_016873262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832233 | Chr.11:94248268 - 94425305 on Build GRCh38 | Gain |
|
| esv2671948 | Chr.11:94349534 - 94645424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map