Assay Details
Target Gene Details
Entrez Gene ID: | 6947 |
Gene Name: | transcobalamin 1 |
Gene Aliases: |
HC, TC-1, TC1, TCI |
Location: |
Chr.11:59852808-59866568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCN1 | NM_001062.3 | NP_001053.2 | ||
| AK292990.1 | ||||
| J05068.1 | AAA61058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626542 | Chr.11:59847853 - 59859203 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map