Assay Details
Target Gene Details
Entrez Gene ID: | 23244 |
Gene Name: | PDS5 cohesin associated factor A |
Gene Aliases: |
PIG54, SCC-112, SCC112 |
Location: |
Chr.4:39822863-39977956 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDS5A | NM_001100399.1 | 33 | 5522 | NP_001093869.1 |
| XM_011513672.2 | 33 | 5135 | XP_011511974.1 | |
| XM_017007928.1 | 33 | 5097 | XP_016863417.1 | |
| AB014548.1 | 21 | 3526 | BAA31623.1 | |
| AB621803.1 | 33 | 5536 | ||
| AF294791.1 | 33 | 5093 | AAM82347.1 | |
| AK001828.1 | 1 | 349 | ||
| AK021757.1 | 8 | 2008 | ||
| BC009650.1 | 9 | 2107 | ||
| BC114218.1 | 33 | 5218 | AAI14219.1 | |
| CR749425.1 | 32 | 4891 | CAH18263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594030 | Chr.4:39767966 - 39835508 on Build GRCh38 | Gain |
|
| nsv594031 | Chr.4:39808834 - 39856900 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map