Assay Details
Target Gene Details
Entrez Gene ID: | 58504 |
Gene Name: | Rho GTPase activating protein 22 |
Gene Aliases: |
RhoGAP2, RhoGap22 |
Location: |
Chr.10:48429831-48656265 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP22 | XM_011540013.2 | XP_011538315.1 | ||
| XM_017016473.1 | XP_016871962.1 | |||
| XM_017016476.1 | XP_016871965.1 |
Target Gene Details
Entrez Gene ID: | 5599 |
Gene Name: | mitogen-activated protein kinase 8 |
Gene Aliases: |
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c |
Location: |
Chr.10:48306639-48439360 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518744 | Chr.10:48428464 - 48435183 on Build GRCh38 | Loss |
|
| nsv1039508 | Chr.10:48387675 - 48446360 on Build GRCh38 | Gain |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map