Assay Details
Target Gene Details
Entrez Gene ID: | 2041 |
Gene Name: | EPH receptor A1 |
Gene Aliases: |
EPH, EPHT, EPHT1 |
Location: |
Chr.7:143390813-143408892 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA1 | XM_006715880.3 | 17 | 3081 | XP_006715943.1 |
Target Gene Details
Entrez Gene ID: | 7791 |
Gene Name: | zyxin |
Gene Aliases: |
ESP-2, HED-2 |
Location: |
Chr.7:143381174-143391111 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZYX | NM_001010972.1 | 10 | 2214 | NP_001010972.1 |
| NM_003461.4 | 10 | 2217 | NP_003452.1 | |
| XM_011516569.2 | 9 | 2112 | XP_011514871.2 | |
| XM_017012587.1 | 10 | 2307 | XP_016868076.1 | |
| AK130138.1 | 6 | 1507 | ||
| AK298925.1 | 8 | 1963 | ||
| AK299005.1 | 9 | 1933 | ||
| BC002323.2 | 10 | 2086 | AAH02323.2 | |
| BC008743.2 | 10 | 2181 | AAH08743.1 | |
| BC009360.2 | 10 | 2126 | AAH09360.1 | |
| BC010031.2 | 10 | 2114 | AAH10031.1 | |
| BC017183.2 | 9 | 1743 | AAH17183.2 | |
| U15158.1 | 8 | 1822 | AAA78902.1 | |
| X94991.1 | 9 | 2069 | CAA64447.1 | |
| X95735.1 | 10 | 2130 | CAA65050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949967 | Chr.7:143382508 - 143399907 on Build GRCh38 | Deletion |
|
| nsv482115 | Chr.7:143381267 - 143391109 on Build GRCh38 | Gain |
|
| nsv608681 | Chr.7:143372217 - 143425364 on Build GRCh38 | Loss |
|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map