Assay Details
Target Gene Details
Entrez Gene ID: | 10020 |
Gene Name: | glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase |
Gene Aliases: |
DMRV, GLCNE, IBM2, NM, Uae1 |
Location: |
Chr.9:36214441-36277056 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNE | NM_001128227.2 | NP_001121699.1 | ||
| NM_001190383.1 | NP_001177312.1 | |||
| NM_001190384.1 | NP_001177313.1 | |||
| NM_001190388.1 | NP_001177317.1 | |||
| NM_005476.5 | NP_005467.1 | |||
| XM_005251334.4 | XP_005251391.1 | |||
| XM_017014167.1 | XP_016869656.1 | |||
| XM_017014168.1 | XP_016869657.1 | |||
| AF051852.1 | AAD32251.1 | |||
| AF155663.1 | AAD38197.1 | |||
| AJ238764.1 | CAB42607.1 | |||
| AK295562.1 | ||||
| AK296687.1 | ||||
| AK299488.1 | ||||
| AK312539.1 | ||||
| AM697708.1 | ||||
| AM697709.1 | ||||
| AY531126.1 | AAS59257.1 | |||
| AY531127.1 | AAS59258.1 | |||
| AY531128.1 | AAS59259.1 | |||
| BC121179.2 | ||||
| EU093084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017711 | Chr.9:36073424 - 36548245 on Build GRCh38 | Gain |
|
| esv34080 | Chr.9:36005887 - 36453179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map