Assay Details
Target Gene Details
Entrez Gene ID: | 3558 |
Gene Name: | interleukin 2 |
Gene Aliases: |
IL-2, TCGF, lymphokine |
Location: |
Chr.4:122449479-122456495 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL2 | NM_000586.3 | NP_000577.2 | ||
| XM_017008177.1 | XP_016863666.1 | |||
| A14844.1 | CAA01199.1 | |||
| AB451325.1 | ||||
| AB451454.1 | ||||
| AF228636.1 | AAG53575.1 | |||
| AF532913.1 | AAM97916.1 | |||
| AY283686.1 | AAP35033.1 | |||
| AY523040.1 | AAS17753.1 | |||
| BC066254.1 | AAH66254.1 | |||
| BC066255.1 | AAH66255.1 | |||
| BC066256.1 | AAH66256.1 | |||
| BC066257.1 | AAH66257.1 | |||
| BC070338.1 | AAH70338.1 | |||
| DQ231169.1 | ABB20900.1 | |||
| DQ861285.1 | ||||
| S77834.1 | AAD14263.2 | |||
| S77835.1 | AAD14264.1 | |||
| S82692.1 | AAB46883.1 | |||
| U25676.1 | AAA70092.1 | |||
| V00564.1 | CAA23827.1 | |||
| X01586.1 | CAA25742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894021 | Chr.4:122306958 - 122473886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map