Assay Details
Target Gene Details
Entrez Gene ID: | 9381 |
Gene Name: | otoferlin |
Gene Aliases: |
AUNB1, DFNB6, DFNB9, FER1L2, NSRD9 |
Location: |
Chr.2:26457203-26558698 on Build GRCh38 |
Assay Gene Location: | Within Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTOF | NM_001287489.1 | NP_001274418.1 | ||
| NM_004802.3 | 29 | 3836 | NP_004793.2 | |
| NM_194248.2 | 46 | 6038 | NP_919224.1 | |
| NM_194322.2 | 28 | 3990 | NP_919303.1 | |
| NM_194323.2 | NP_919304.1 | |||
| XM_017005338.1 | 46 | 5851 | XP_016860827.1 | |
| AF107403.1 | 29 | 3836 | AAD26117.1 | |
| AF183185.1 | 46 | 6038 | AAG12991.1 | |
| AF183186.1 | 28 | 3990 | AAG12992.1 | |
| AF183187.1 | AAG17468.1 | |||
| AK296272.1 | ||||
| EU436746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583675 | Chr.2:26440850 - 26577652 on Build GRCh38 | Loss |
|
| nsv517800 | Chr.2:26449146 - 26617624 on Build GRCh38 | Loss |
|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
| nsv953759 | Chr.2:26459433 - 26484332 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map