Assay Details
Target Gene Details
Entrez Gene ID: | 11321 |
Gene Name: | GPN-loop GTPase 1 |
Gene Aliases: |
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1 |
Location: |
Chr.2:27628648-27650846 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPN1 | NM_001145047.1 | NP_001138519.1 | ||
| NM_001145048.1 | NP_001138520.1 | |||
| NM_001145049.1 | NP_001138521.1 | |||
| NM_007266.3 | NP_009197.2 | |||
| NR_026735.1 | ||||
| AB044661.1 | BAB17612.1 | |||
| AJ010842.1 | CAA09376.1 | |||
| AK298827.1 | ||||
| AK298866.1 | ||||
| AK302131.1 | ||||
| AK310340.1 | ||||
| AK311423.1 | ||||
| AK312992.1 | ||||
| AK316404.1 | ||||
| BC007451.2 | AAH07451.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79898842] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map