Assay Details
Target Gene Details
Entrez Gene ID: | 57469 |
Gene Name: | paraneoplastic Ma antigen family-like 2 |
Gene Aliases: |
- |
Location: |
Chr.19:46491191-46495912 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNMAL2 | NM_020709.2 | 1 | 601 | NP_065760.1 |
| AK124720.1 | 1 | 601 | BAC85931.1 | |
| DA015488.1 |
Target Gene Details
Entrez Gene ID: | 100506012 |
Gene Name: | PPP5 tetratricopeptide repeat domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.19:46480788-46601200 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP5D1 | NM_001205281.1 | NP_001192210.1 | ||
| BM718195.1 |
More Information
Set Membership: |
|

Genomic Map