Assay Details
Target Gene Details
Entrez Gene ID: | 79685 |
Gene Name: | SAP30 like |
Gene Aliases: |
NS4ATP2 |
Location: |
Chr.5:154445957-154461053 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP30L | NM_001131062.1 | NP_001124534.1 | ||
| NM_001131063.1 | NP_001124535.1 | |||
| NM_024632.5 | NP_078908.1 | |||
| NR_024084.1 | ||||
| AI144341.1 | ||||
| AI199517.1 | ||||
| AI436556.1 | ||||
| AK021588.1 | BAB13848.1 | |||
| AY341060.1 | AAQ16562.1 | |||
| AY846876.1 | AAX54477.1 | |||
| BC009829.2 | AAH09829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830520 | Chr.5:154372285 - 154553755 on Build GRCh38 | Loss |
|
| nsv1015362 | Chr.5:154215521 - 154450098 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map