Assay Details
Target Gene Details
Entrez Gene ID: | 2797 |
Gene Name: | gonadotropin releasing hormone 2 |
Gene Aliases: |
GnRH-II, LH-RHII |
Location: |
Chr.20:3039061-3045896 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNRH2 | NM_001310220.1 | NP_001297149.1 | ||
| NM_001501.1 | NP_001492.1 | |||
| NM_178331.1 | NP_847901.1 | |||
| NM_178332.1 | NP_847902.1 | |||
| XM_017027823.1 | XP_016883312.1 | |||
| AF036330.1 | AAC02981.1 | |||
| AF190640.1 | AAG17539.1 | |||
| BC069362.1 | AAH69362.1 | |||
| BC115399.1 | ||||
| BC115400.1 | ||||
| BF061974.1 | ||||
| BF062277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057187 | Chr.20:2906193 - 3065597 on Build GRCh38 | Gain |
|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833896 | Chr.20:2923851 - 3062392 on Build GRCh38 | Loss |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
| nsv482064 | Chr.20:3043622 - 3045745 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map