Assay Details
Target Gene Details
Entrez Gene ID: | 84557 |
Gene Name: | microtubule associated protein 1 light chain 3 alpha |
Gene Aliases: |
ATG8E, LC3, LC3A, MAP1ALC3, MAP1BLC3 |
Location: |
Chr.20:34546823-34560345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP1LC3A | NM_032514.3 | NP_115903.1 | ||
| NM_181509.2 | NP_852610.1 | |||
| XM_011529083.2 | XP_011527385.1 | |||
| XM_011529084.2 | XP_011527386.1 | |||
| XM_011529085.2 | XP_011527387.1 | |||
| AF276658.1 | AAK35151.1 | |||
| AL833855.1 | CAD38714.1 | |||
| BC015810.1 | AAH15810.1 | |||
| BM740344.1 | ||||
| BP212418.1 | ||||
| BT007452.1 | AAP36120.1 | |||
| EL954540.1 | ||||
| H18312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3359 | Chr.20:34531574 - 34576232 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map