Assay Details
Target Gene Details
Entrez Gene ID: | 137868 |
Gene Name: | sarcoglycan zeta |
Gene Aliases: |
ZSG1 |
Location: |
Chr.8:14084835-15238283 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGCZ | NM_001322879.1 | 7 | 1862 | NP_001309808.1 |
| NM_001322880.1 | 7 | 1841 | NP_001309809.1 | |
| NM_001322881.1 | 7 | 1747 | NP_001309810.1 | |
| NM_139167.3 | 8 | 1964 | NP_631906.2 | |
| AY028700.1 | 7 | 1265 | AAK21962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616348 | Chr.8:13970635 - 14105055 on Build GRCh38 | Loss |
|
| nsv465524 | Chr.8:14037682 - 14115956 on Build GRCh38 | Gain |
|
| nsv517019 | Chr.8:13511552 - 14801106 on Build GRCh38 | Gain+Loss |
|
| dgv7101n100 | Chr.8:14038545 - 14109037 on Build GRCh38 | Gain |
|
| nsv470190 | Chr.8:14037681 - 14115956 on Build GRCh38 | Gain |
|
| nsv610452 | Chr.8:13799452 - 14103624 on Build GRCh38 | Loss |
|
| esv3616353 | Chr.8:14044616 - 14186950 on Build GRCh38 | Gain |
|
| nsv610411 | Chr.8:13093516 - 14267300 on Build GRCh38 | Loss |
|
| nsv1022834 | Chr.8:13067043 - 14121780 on Build GRCh38 | Gain |
|
| nsv1031710 | Chr.8:13987713 - 14121675 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map