Assay Details
Target Gene Details
Entrez Gene ID: | 160418 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 3 |
Gene Aliases: |
SMILE |
Location: |
Chr.12:88142296-88199887 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMTC3 | NM_181783.3 | 2 | 267 | NP_861448.2 |
| XM_011537980.2 | 2 | 261 | XP_011536282.1 | |
| AJ697717.1 | 2 | 137 | CAG26973.1 | |
| AK290436.1 | 2 | 267 | ||
| AK302755.1 | 2 | 267 | ||
| AK310375.1 | 2 | 267 | ||
| AK310393.1 | 2 | 267 | ||
| BC117175.1 | 1 | 88 | ||
| BC117177.1 | 1 | 89 | ||
| BX538169.1 | 2 | 240 | CAD98046.1 | |
| CR749244.1 | 2 | 219 | CAH18100.1 | |
| CR749309.1 | 2 | 253 | CAH18164.1 | |
| CR933632.1 | 2 | 218 | CAI45938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv807 | Chr.12:88133831 - 88178716 on Build GRCh38 | Deletion |
|
| nsv483111 | Chr.12:86306223 - 88606223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map