Assay Details
Target Gene Details
Entrez Gene ID: | 25792 |
Gene Name: | CDKN1A interacting zinc finger protein 1 |
Gene Aliases: |
LSFR1, NP94, ZNF356 |
Location: |
Chr.9:128166065-128204383 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIZ1 | NM_001131015.1 | NP_001124487.1 | ||
| NM_012127.2 | NP_036259.2 | |||
| AK023978.1 | BAB14750.1 |
Target Gene Details
Entrez Gene ID: | 1759 |
Gene Name: | dynamin 1 |
Gene Aliases: |
DNM, EIEE31 |
Location: |
Chr.9:128203355-128255248 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNM1 | NM_001005336.2 | 1 | 128 | NP_001005336.1 |
| NM_001288737.1 | 1 | 128 | NP_001275666.1 | |
| NM_001288738.1 | 1 | 128 | NP_001275667.1 | |
| NM_001288739.1 | 1 | 128 | NP_001275668.1 | |
| NM_004408.3 | 1 | 128 | NP_004399.2 | |
| XM_005251763.2 | 1 | 128 | XP_005251820.1 | |
| XM_005251768.2 | 1 | 128 | XP_005251825.1 | |
| XM_005251769.2 | 1 | 128 | XP_005251826.1 | |
| XM_006716992.2 | 1 | 128 | XP_006717055.1 | |
| XM_006716993.1 | 1 | 128 | XP_006717056.1 | |
| XM_011518335.2 | 1 | 128 | XP_011516637.1 | |
| XM_011518336.2 | 1 | 128 | XP_011516638.1 | |
| XM_017014369.1 | 1 | 128 | XP_016869858.1 | |
| XM_017014370.1 | 1 | 128 | XP_016869859.1 | |
| XM_017014371.1 | 1 | 128 | XP_016869860.1 | |
| XM_017014372.1 | 1 | 128 | XP_016869861.1 | |
| XM_017014373.1 | 1 | 128 | XP_016869862.1 | |
| AK225179.1 | ||||
| AK295101.1 | ||||
| AK296326.1 | 1 | 95 | ||
| AK316235.1 | 1 | 104 | ||
| BC050279.1 | 1 | 104 | AAH50279.2 | |
| BC063850.1 | 1 | 61 | AAH63850.1 | |
| DA221568.1 | 1 | 128 | ||
| DA510154.1 | 1 | 108 | ||
| L07807.1 | 1 | 78 | AAA02803.1 |
More Information
Set Membership: |
|

Genomic Map