Assay Details
Target Gene Details
Entrez Gene ID: | 51147 |
Gene Name: | inhibitor of growth family member 4 |
Gene Aliases: |
my036, p29ING4 |
Location: |
Chr.12:6650538-6663143 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ING4 | NM_001127582.1 | NP_001121054.1 | ||
| NM_001127583.1 | NP_001121055.1 | |||
| NM_001127584.1 | NP_001121056.1 | |||
| NM_001127585.1 | NP_001121057.1 | |||
| NM_001127586.1 | NP_001121058.1 | |||
| NM_016162.3 | NP_057246.2 | |||
| XM_005253698.3 | XP_005253755.1 | |||
| XM_011520964.2 | XP_011519266.1 | |||
| XM_011520965.2 | XP_011519267.1 | |||
| XM_017019392.1 | XP_016874881.1 | |||
| XM_017019393.1 | XP_016874882.1 | |||
| AF063594.1 | AAG43153.1 | |||
| AF110645.1 | AAD48585.1 | |||
| AF156552.1 | AAL79773.1 | |||
| AW515338.1 | 1 | 285 | ||
| AW614868.1 | ||||
| BC013038.2 | AAH13038.2 | |||
| BC095434.1 | AAH95434.1 | |||
| BM727982.1 | 1 | 214 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509452 | Chr.12:6566935 - 6653627 on Build GRCh38 | Insertion |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map