Assay Details
Target Gene Details
Entrez Gene ID: | 51528 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
C14orf100, CDA06, HSPC213, HSPC327, JAMP |
Location: |
Chr.14:59484443-59505407 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JKAMP | NM_001098625.2 | 7 | 917 | NP_001092095.1 |
| NM_001284201.1 | 7 | 1458 | NP_001271130.1 | |
| NM_001284202.1 | 8 | 1044 | NP_001271131.1 | |
| NM_001284203.1 | 7 | 967 | NP_001271132.1 | |
| NM_001284204.1 | 6 | 858 | NP_001271133.1 | |
| NM_016475.4 | 7 | 935 | NP_057559.2 | |
| XM_006720171.3 | 6 | 834 | XP_006720234.1 | |
| XM_017021366.1 | 6 | 767 | XP_016876855.1 | |
| AF151047.1 | 7 | 857 | AAF36133.1 | |
| AF161445.1 | 7 | 856 | AAF29005.1 | |
| AF212245.1 | 7 | 857 | AAG41781.1 | |
| AF529304.1 | 5 | 1172 | AAP85636.1 | |
| AK021683.1 | 7 | 816 | BAB13874.1 | |
| AK027591.1 | 7 | 861 | BAB55216.1 | |
| AK056145.1 | 7 | 1334 | ||
| AK128628.1 | 1 | 2741 | ||
| AK298209.1 | 7 | 842 | ||
| AL136696.1 | 7 | 827 | CAB66631.1 | |
| BC005198.1 | 7 | 843 | AAH05198.1 | |
| BC010359.1 | 7 | 874 | AAH10359.1 | |
| BU928810.1 | 6 | 785 | ||
| BX161501.1 | 8 | 927 | CAD61945.1 | |
| CR457163.1 | 6 | 770 | CAG33444.1 | |
| CR533510.1 | 6 | 788 | CAG38541.1 |
More Information
Set Membership: |
|

Genomic Map