Assay Details
Target Gene Details
Entrez Gene ID: | 79048 |
Gene Name: | SECIS binding protein 2 |
Gene Aliases: |
SBP2 |
Location: |
Chr.9:89318473-89367117 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SECISBP2 | NM_001282688.1 | 11 | 1613 | NP_001269617.1 |
| NM_001282689.1 | 11 | 1495 | NP_001269618.1 | |
| NM_001282690.1 | 11 | 1693 | NP_001269619.1 | |
| NM_024077.4 | 11 | 1616 | NP_076982.3 | |
| XM_005252196.2 | 11 | 1466 | XP_005252253.1 | |
| XM_005252202.3 | 7 | 848 | XP_005252259.1 | |
| XM_006717282.2 | 11 | 1549 | XP_006717345.1 | |
| XM_011519000.2 | 11 | 1670 | XP_011517302.1 | |
| XM_011519001.1 | 10 | 1580 | XP_011517303.1 | |
| XM_011519002.1 | 11 | 1556 | XP_011517304.1 | |
| XM_011519003.1 | 11 | 1553 | XP_011517305.1 | |
| XM_017015122.1 | 10 | 1493 | XP_016870611.1 | |
| XM_017015123.1 | 11 | 1469 | XP_016870612.1 | |
| XM_017015124.1 | 10 | 1376 | XP_016870613.1 | |
| XM_017015125.1 | 9 | 1171 | XP_016870614.1 | |
| AB208940.1 | 11 | 1536 | BAD92177.1 | |
| AF380995.1 | 11 | 1569 | AAK57518.1 | |
| AK023078.1 | 4 | 347 | BAB14393.1 | |
| AK090608.1 | 11 | 1583 | ||
| AK290182.1 | 11 | 1462 | ||
| AK299161.1 | 11 | 1704 | ||
| AK302852.1 | 11 | 1611 | ||
| AL136881.1 | 11 | 1449 | CAB66815.1 | |
| BC023142.1 | 5 | 463 | AAH23142.2 | |
| BC036109.1 | 11 | 1578 | AAH36109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| nsv1041327 | Chr.9:89298100 - 89438209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map