Assay Details
Target Gene Details
Entrez Gene ID: | 196740 |
Gene Name: | V-set and transmembrane domain containing 4 |
Gene Aliases: |
C10orf72 |
Location: |
Chr.10:49014245-49129118 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM4 | NM_001031746.4 | 8 | 1492 | NP_001026916.2 |
| XM_017015828.1 | 8 | 1912 | XP_016871317.1 | |
| BC041414.1 | 8 | 1474 | AAH41414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623162 | Chr.10:49011836 - 49038527 on Build GRCh38 | Loss |
|
| nsv831866 | Chr.10:48957135 - 49127231 on Build GRCh38 | Gain+Loss |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map