Assay Details
Target Gene Details
Entrez Gene ID: | 101928140 |
Gene Name: | uncharacterized LOC101928140 |
Gene Aliases: |
- |
Location: |
Chr.6:128027865-128086293 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928140 | NR_125849.1 |
Target Gene Details
Entrez Gene ID: | 5796 |
Gene Name: | protein tyrosine phosphatase, receptor type K |
Gene Aliases: |
R-PTP-kappa |
Location: |
Chr.6:127968779-128520847 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604643 | Chr.6:128038217 - 128172846 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map