Assay Details
Target Gene Details
Entrez Gene ID: | 3684 |
Gene Name: | integrin subunit alpha M |
Gene Aliases: |
CD11B, CR3A, MAC-1, MAC1A, MO1A, SLEB6 |
Location: |
Chr.16:31259967-31332892 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGAM | NM_000632.3 | 6 | 594 | NP_000623.2 |
| NM_001145808.1 | 6 | 594 | NP_001139280.1 | |
| XM_006721045.1 | 6 | 586 | XP_006721108.1 | |
| XM_011545850.2 | 4 | 390 | XP_011544152.1 | |
| XM_011545851.2 | 6 | 586 | XP_011544153.1 | |
| XM_017023216.1 | 6 | 586 | XP_016878705.1 | |
| AK057856.1 | ||||
| AK127637.1 | ||||
| BC096346.3 | 6 | 560 | AAH96346.1 | |
| BC096347.3 | 6 | 560 | AAH96347.1 | |
| BC096348.3 | 6 | 560 | AAH96348.1 | |
| BC099660.3 | 6 | 560 | AAH99660.1 | |
| J03925.1 | 6 | 568 | AAA59544.1 | |
| J04145.1 | 5 | 472 | AAA59903.1 | |
| M18044.1 | 6 | 571 | AAA59491.1 |
More Information
Set Membership: |
|

Genomic Map