Assay Details
Target Gene Details
Entrez Gene ID: | 253017 |
Gene Name: | trans-2,3-enoyl-CoA reductase-like |
Gene Aliases: |
GPSN2L, SRD5A2L2, TERL |
Location: |
Chr.4:64276298-64409509 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TECRL | NM_001010874.4 | 10 | 991 | NP_001010874.2 |
| XM_005265662.4 | 9 | 995 | XP_005265719.1 | |
| XM_005265663.2 | 10 | 991 | XP_005265720.1 | |
| XM_005265664.2 | 10 | 991 | XP_005265721.1 | |
| XM_005265665.3 | 10 | 991 | XP_005265722.1 | |
| XM_017007960.1 | XP_016863449.1 | |||
| AK290606.1 | 10 | 987 | ||
| AL833108.1 | 10 | 992 | CAI46109.1 | |
| BX647590.1 | 10 | 928 | CAI46063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998669 | Chr.4:63850701 - 64580449 on Build GRCh38 | Loss |
|
| nsv1012062 | Chr.4:64277750 - 64318606 on Build GRCh38 | Loss |
|
| nsv594372 | Chr.4:62968251 - 65067958 on Build GRCh38 | Loss |
|
| nsv594411 | Chr.4:63409748 - 64304697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map