Assay Details
Target Gene Details
Entrez Gene ID: | 171389 |
Gene Name: | NLR family pyrin domain containing 6 |
Gene Aliases: |
AVR, CLR11.4, NALP6, NAVR, NAVR/AVR, PAN3, PYPAF5 |
Location: |
Chr.11:278365-285942 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP6 | NM_001276700.1 | 8 | 2650 | NP_001263629.1 |
| NM_138329.2 | 8 | 2653 | NP_612202.2 | |
| XM_017017253.1 | 5 | 2337 | XP_016872742.1 | |
| AF479748.1 | AAL87105.1 | |||
| AK292668.1 | 8 | 2855 | ||
| AY154461.1 | AAO18157.1 | |||
| CD636365.1 | 1 | 111 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32593 | Chr.11:218394 - 285937 on Build GRCh38 | Loss |
|
| esv3891905 | Chr.11:269147 - 336332 on Build GRCh38 | Gain |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1012n100 | Chr.11:198510 - 427836 on Build GRCh38 | Gain |
|
| nsv552766 | Chr.11:248181 - 285936 on Build GRCh38 | Loss |
|
| nsv482153 | Chr.11:278570 - 285303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map