Assay Details
Target Gene Details
Entrez Gene ID: | 54360 |
Gene Name: | cytokine like 1 |
Gene Aliases: |
C17, C4orf4 |
Location: |
Chr.4:5014586-5019470 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYTL1 | NM_018659.2 | 4 | 883 | NP_061129.1 |
| XM_017008299.1 | 5 | 998 | XP_016863788.1 | |
| AF193766.1 | 4 | 886 | AAF73372.1 | |
| AF274956.1 | 4 | 743 | AAK07531.1 | |
| AY359101.1 | 4 | 884 | AAQ89459.1 | |
| BC031391.1 | 4 | 859 | AAH31391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5094n100 | Chr.4:4920733 - 5083483 on Build GRCh38 | Gain |
|
| nsv593479 | Chr.4:4899358 - 5459331 on Build GRCh38 | Gain |
|
| nsv997889 | Chr.4:4785891 - 5220458 on Build GRCh38 | Gain |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map