Assay Details
Target Gene Details
Entrez Gene ID: | 84164 |
Gene Name: | activating signal cointegrator 1 complex subunit 2 |
Gene Aliases: |
ASC1p100, p100 |
Location: |
Chr.22:29788608-29838304 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3590 | Chr.22:29770034 - 29790199 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map