Assay Details
Target Gene Details
Entrez Gene ID: | 51379 |
Gene Name: | cytokine receptor like factor 3 |
Gene Aliases: |
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2 |
Location: |
Chr.17:30782684-30824760 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRLF3 | NM_015986.3 | 7 | 1121 | NP_057070.3 |
| NR_073118.1 | 6 | 913 | ||
| XM_011524879.1 | 7 | 1295 | XP_011523181.1 | |
| AF046059.1 | 7 | 1034 | AAD02422.1 | |
| AF120151.1 | 6 | 1487 | AAD31758.1 | |
| AK296238.1 | 6 | 824 | ||
| AK313315.1 | 7 | 1110 | ||
| BC023567.2 | 7 | 1043 | AAH23567.2 | |
| DQ298450.1 | 7 | 1000 | ABC55264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892992 | Chr.17:30769803 - 30860564 on Build GRCh38 | Loss |
|
| nsv457720 | Chr.17:30625268 - 31022670 on Build GRCh38 | Loss |
|
| nsv833411 | Chr.17:30739376 - 30792370 on Build GRCh38 | Loss |
|
| esv2715813 | Chr.17:30601472 - 31009462 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map