Assay Details
Target Gene Details
Entrez Gene ID: | 8322 |
Gene Name: | frizzled class receptor 4 |
Gene Aliases: |
CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4 |
Location: |
Chr.11:86945675-86955398 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FZD4 | NM_012193.3 | 2 | 2929 | NP_036325.2 |
| AB032417.1 | 2 | 2922 | BAA86286.1 | |
| BC114527.1 | 2 | 2790 | ||
| BC114622.1 | 2 | 2790 |
Target Gene Details
Entrez Gene ID: | 11098 |
Gene Name: | protease, serine 23 |
Gene Aliases: |
SIG13, SPUVE, ZSIG13 |
Location: |
Chr.11:86791059-86952910 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS23 | NR_120591.1 | |||
| NR_120592.1 | ||||
| AU279649.1 | ||||
| DA396057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627099 | Chr.11:86660723 - 87045565 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map