Assay Details
Target Gene Details
Entrez Gene ID: | 54896 |
Gene Name: | PQ loop repeat containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:19312246-19332611 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PQLC2 | NM_001040125.1 | 8 | 1726 | NP_001035214.1 |
| NM_001040126.1 | 8 | 1114 | NP_001035215.1 | |
| NM_001287531.1 | 8 | 1083 | NP_001274460.1 | |
| NM_017765.2 | 9 | 1356 | NP_060235.2 | |
| NR_109848.1 | 9 | 1356 | ||
| XM_005245916.2 | 8 | 1139 | XP_005245973.1 | |
| XM_006710710.2 | 9 | 1249 | XP_006710773.1 | |
| XM_006710711.2 | 8 | 1108 | XP_006710774.1 | |
| XM_011541634.2 | 8 | 2123 | XP_011539936.1 | |
| AK000327.1 | 9 | 1277 | BAA91088.1 | |
| AK125580.1 | 8 | 1644 | ||
| AK130171.1 | 9 | 1274 | BAC85296.1 | |
| AK130175.1 | 9 | 1276 | BAC85298.1 | |
| AK172745.1 | 8 | 1001 | BAD18732.1 | |
| BC015324.1 | 8 | 1019 | AAH15324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map