Assay Details
Target Gene Details
Entrez Gene ID: | 6434 |
Gene Name: | transformer 2 beta homolog (Drosophila) |
Gene Aliases: |
Htra2-beta, PPP1R156, SFRS10, SRFS10, TRA2-BETA, TRAN2B |
Location: |
Chr.3:185914568-185938136 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRA2B | NM_001243879.1 | 2 | 433 | NP_001230808.1 |
| NM_004593.2 | 3 | 567 | NP_004584.1 | |
| XM_005247703.1 | 4 | 735 | XP_005247760.1 | |
| XM_011513072.2 | 4 | 3537 | XP_011511374.1 | |
| XM_017007033.1 | 3 | 1713 | XP_016862522.1 | |
| AB209208.1 | 4 | 3524 | BAD92445.1 | |
| AK098191.1 | 3 | 354 | BAC05256.1 | |
| AK301118.1 | 2 | 286 | ||
| AK308267.1 | 2 | 311 | ||
| AK315632.1 | 3 | 551 | ||
| BC000160.2 | 3 | 434 | AAH00160.1 | |
| BC000451.2 | 3 | 433 | AAH00451.1 | |
| BC005898.1 | 3 | 418 | AAH05898.1 | |
| CR749214.1 | 2 | 2594 | CAH18071.1 | |
| U61267.1 | 3 | 412 | AAC28242.1 | |
| U68063.1 | 3 | 420 | AAB08701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598980 | Chr.3:185910936 - 185991411 on Build GRCh38 | Gain |
|
| nsv1142591 | Chr.3:185911072 - 185991507 on Build GRCh38 | Deletion |
|
| esv2660289 | Chr.3:185910992 - 185991468 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map